N-Phenethyl-4-phenoxyaniline

Supplier: Matrix Scientific

031453-500MG
101831-732EA 306.15 USD
101831-732
N-Phenethyl-4-phenoxyaniline
N-Phenethyl-4-phenoxyaniline

Hazard: Irritant

Formula: C₂₀H₁₉NO
MDL Number: MFCD06408201
CAS Number: 1024240-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR