N-(3-Ethoxybenzyl)-4-fluoroaniline

Supplier: Matrix Scientific

031447-500MG
101831-720EA 306.15 USD
101831-720
N-(3-Ethoxybenzyl)-4-fluoroaniline
N-(3-Ethoxybenzyl)-4-fluoroaniline

Hazard: Irritant

Formula: C₁₅H₁₆FNO
MDL Number: MFCD04323440
CAS Number: 801242-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR