4-Fluoro-N-(4-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

031444-500MG
101831-714EA 306.15 USD
101831-714
4-Fluoro-N-(4-isopropoxybenzyl)aniline
4-Fluoro-N-(4-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₈FNO
MDL Number: MFCD10687551
CAS Number: 1040687-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR