4-Fluoro-N-(2-propoxybenzyl)aniline

Supplier: Matrix Scientific

031443-500MG
101831-712EA 306.15 USD
101831-712
4-Fluoro-N-(2-propoxybenzyl)aniline
4-Fluoro-N-(2-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₈FNO
MDL Number: MFCD10687550
CAS Number: 1040687-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR