4-Fluoro-N-(4-isobutoxybenzyl)aniline

Supplier: Matrix Scientific

031442-500MG
101831-710EA 306.15 USD
101831-710
4-Fluoro-N-(4-isobutoxybenzyl)aniline
4-Fluoro-N-(4-isobutoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₀FNO
MDL Number: MFCD10687549
CAS Number: 1040687-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR