N-(2-Thienylmethyl)-1-hexadecanamine

Supplier: Matrix Scientific

031441-500MG
101831-708EA 306.15 USD
101831-708
N-(2-Thienylmethyl)-1-hexadecanamine
N-(2-Thienylmethyl)-1-hexadecanamine

Hazard: Irritant

Formula: C₂₁H₃₉NS
MDL Number: MFCD10687548
CAS Number: 855956-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR