N-[2-(4-Ethylphenoxy)ethyl]-4-fluoroaniline

Supplier: Matrix Scientific

031440-500MG
101831-706EA 306.15 USD
101831-706
N-[2-(4-Ethylphenoxy)ethyl]-4-fluoroaniline
N-[2-(4-Ethylphenoxy)ethyl]-4-fluoroaniline

Hazard: Irritant

Formula: C₁₆H₁₈FNO
MDL Number: MFCD10687547
CAS Number: 1040687-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR