N-[2-(2,5-Dimethylphenoxy)ethyl]-4-fluoroaniline

Supplier: Matrix Scientific

031439-500MG
101831-704EA 306.15 USD
101831-704
N-[2-(2,5-Dimethylphenoxy)ethyl]-4-fluoroaniline
N-[2-(2,5-Dimethylphenoxy)ethyl]-4-fluoroaniline

Hazard: Irritant

Formula: C₁₆H₁₈FNO
MDL Number: MFCD10687546
CAS Number: 1040687-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR