N-(2-Chlorobenzyl)-1-naphthalenamine
Supplier: Matrix Scientific
031438-500MG
101831-702EA
306.15
USD
101831-702
N-(2-Chlorobenzyl)-1-naphthalenamine
N-(2-Chlorobenzyl)-1-naphthalenamine
Hazard: Irritant
Formula:
C₁₇H₁₄ClN |
MDL Number:
MFCD03210812 CAS Number: 113250-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...