4-Fluoro-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

031436-500MG
101831-698EA 306.15 USD
101831-698
4-Fluoro-N-(3-methylbenzyl)aniline
4-Fluoro-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₄FN
MDL Number: MFCD03210619
CAS Number: 356530-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR