N-(3-Chlorobenzyl)-4-ethoxyaniline

Supplier: Matrix Scientific

031431-500MG
101831-688EA 306.15 USD
101831-688
N-(3-Chlorobenzyl)-4-ethoxyaniline
N-(3-Chlorobenzyl)-4-ethoxyaniline

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD06408096
CAS Number: 1019567-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR