4-Ethoxy-N-[2-(3-methylphenoxy)propyl]aniline

Supplier: Matrix Scientific

031430-500MG
101831-686EA 306.15 USD
101831-686
4-Ethoxy-N-[2-(3-methylphenoxy)propyl]aniline
4-Ethoxy-N-[2-(3-methylphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10687541
CAS Number: 1040687-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR