N-(2,4-Dichlorobenzyl)-1-naphthalenamine

Supplier: Matrix Scientific

031429-500MG
101831-684EA 306.15 USD
101831-684
N-(2,4-Dichlorobenzyl)-1-naphthalenamine
N-(2,4-Dichlorobenzyl)-1-naphthalenamine

Hazard: Irritant

Formula: C₁₇H₁₃Cl₂N
MDL Number: MFCD03210783
CAS Number: 356531-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR