N-[4-(tert-Butyl)benzyl]-4-ethoxyaniline

Supplier: Matrix Scientific

031428-500MG
101831-682EA 306.15 USD
101831-682
N-[4-(tert-Butyl)benzyl]-4-ethoxyaniline
N-[4-(tert-Butyl)benzyl]-4-ethoxyaniline

Hazard: Irritant

Formula: C₁₉H₂₅NO
MDL Number: MFCD10687540
CAS Number: 1002652-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR