N-([1,1'-Biphenyl]-4-ylmethyl)-4-ethoxyaniline

Supplier: Matrix Scientific

031424-500MG
101831-674EA 306.15 USD
101831-674
N-([1,1'-Biphenyl]-4-ylmethyl)-4-ethoxyaniline
N-([1,1'-Biphenyl]-4-ylmethyl)-4-ethoxyaniline

Hazard: Irritant

Formula: C₂₁H₂₁NO
MDL Number: MFCD03210586
CAS Number: 356530-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR