3-Chloro-N,2-dimethylaniline

Supplier: Matrix Scientific

031417-500MG
101831-660EA 306.15 USD
101831-660
3-Chloro-N,2-dimethylaniline
3-Chloro-N,2-dimethylaniline

Hazard: Irritant

Formula: C₈H₁₀ClN
MDL Number: MFCD10687534
CAS Number: 41456-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR