Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(3-Isopropoxybenzyl)(4-methoxyphenyl)methanamine

Supplier: Matrix Scientific


031414-500MG
101831-654EA 260.23 USD InStock
101831-654
N-(3-Isopropoxybenzyl)(4-methoxyphenyl)methanamine
N-(3-Isopropoxybenzyl)(4-methoxyphenyl)methanamine
CAS: 1040686-65-4
MDL Number: MFCD10687532
Molecular Formula: C18H23NO2
Molecular Weight: 285,39
Hazard: Irritant


Formula: C₁₈H₂₃NO₂
MW: 285,39 g/mol
MDL Number: MFCD10687532
CAS Number: 1040686-65-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty