[2-(2-Ethoxyethoxy)phenyl]-N-(4-methoxybenzyl)methanamine

Supplier: Matrix Scientific

031412-500MG
101831-650EA 306.15 USD
101831-650
[2-(2-Ethoxyethoxy)phenyl]-N-(4-methoxybenzyl)methanamine
[2-(2-Ethoxyethoxy)phenyl]-N-(4-methoxybenzyl)methanamine

Hazard: Irritant

Formula: C₁₉H₂₅NO₃
MDL Number: MFCD10687530
CAS Number: 1040686-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR