N-[4-(Heptyloxy)benzyl](4-methoxyphenyl)methanamine

Supplier: Matrix Scientific

031410-500MG
101831-646EA 306.15 USD
101831-646
N-[4-(Heptyloxy)benzyl](4-methoxyphenyl)methanamine
N-[4-(Heptyloxy)benzyl](4-methoxyphenyl)methanamine

Hazard: Irritant

Formula: C₂₂H₃₁NO₂
MW: 341.5 g/mol
MDL Number: MFCD10687529
CAS Number: 1040686-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR