N-(4-Methoxybenzyl)-2-(2-methylphenoxy)-1-propanamine

Supplier: Matrix Scientific

031408-500MG
101831-642EA 306.15 USD
101831-642
N-(4-Methoxybenzyl)-2-(2-methylphenoxy)-1-propanamine
N-(4-Methoxybenzyl)-2-(2-methylphenoxy)-1-propanamine

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10687527
CAS Number: 1040686-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR