N1,N1-Diethyl-N2-[4-(2-methoxyethoxy)benzyl]-1,2-ethanediamine

Supplier: Matrix Scientific

031403-500MG
101831-632EA 306.15 USD
101831-632
N1,N1-Diethyl-N2-[4-(2-methoxyethoxy)benzyl]-1,2-ethanediamine
N1,N1-Diethyl-N2-[4-(2-methoxyethoxy)benzyl]-1,2-ethanediamine

Hazard: Irritant

Formula: C₁₆H₂₈N₂O₂
MDL Number: MFCD10687524
CAS Number: 1040686-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR