N1,N1-Diethyl-N2-[2-(2-methoxyphenoxy)ethyl]-1,2-ethanediamine

Supplier: Matrix Scientific

031398-500MG
101831-622EA 306.15 USD
101831-622
N1,N1-Diethyl-N2-[2-(2-methoxyphenoxy)ethyl]-1,2-ethanediamine
N1,N1-Diethyl-N2-[2-(2-methoxyphenoxy)ethyl]-1,2-ethanediamine

Hazard: Irritant

Formula: C₁₅H₂₆N₂O₂
MDL Number: MFCD10687519
CAS Number: 1226362-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR