N1,N1-Diethyl-N2-phenethyl-1,2-ethanediamine

Supplier: Matrix Scientific

031397-500MG
101831-620EA 306.15 USD
101831-620
N1,N1-Diethyl-N2-phenethyl-1,2-ethanediamine
N1,N1-Diethyl-N2-phenethyl-1,2-ethanediamine

Hazard: Irritant

Formula: C₁₄H₂₄N₂
MDL Number: MFCD08756995
CAS Number: 71108-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR