N-[4-(2-Phenoxyethoxy)benzyl](3-pyridinyl)methanamine

Supplier: Matrix Scientific

031393-500MG
101831-612EA 306.15 USD
101831-612
N-[4-(2-Phenoxyethoxy)benzyl](3-pyridinyl)methanamine
N-[4-(2-Phenoxyethoxy)benzyl](3-pyridinyl)methanamine

Hazard: Irritant

Formula: C₂₁H₂₂N₂O₂
MW: 334.42 g/mol
MDL Number: MFCD10687515
CAS Number: 1040685-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR