2-(2-Methylphenoxy)-N-(3-pyridinylmethyl)-1-ethanamine

Supplier: Matrix Scientific

031387-500MG
101831-600EA 306.15 USD
101831-600
2-(2-Methylphenoxy)-N-(3-pyridinylmethyl)-1-ethanamine
2-(2-Methylphenoxy)-N-(3-pyridinylmethyl)-1-ethanamine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O
MDL Number: MFCD10687509
CAS Number: 1040685-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR