N-(3-Pyridinylmethyl)(2-thienyl)methanamine

Supplier: Matrix Scientific

031386-500MG
101831-598EA 306.15 USD
101831-598
N-(3-Pyridinylmethyl)(2-thienyl)methanamine
N-(3-Pyridinylmethyl)(2-thienyl)methanamine

Hazard: Irritant

Formula: C₁₁H₁₂N₂S
MDL Number: MFCD04530424
CAS Number: 774191-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR