N-(3,5-Dichlorobenzyl)-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

031382-500MG
101831-590EA 306.15 USD
101831-590
N-(3,5-Dichlorobenzyl)-3-(trifluoromethyl)aniline
N-(3,5-Dichlorobenzyl)-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂F₃N
MDL Number: MFCD10687506
CAS Number: 1040685-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR