N-[2-(4-Ethylphenoxy)propyl]-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

031380-500MG
101831-586EA 306.15 USD
101831-586
N-[2-(4-Ethylphenoxy)propyl]-3-(trifluoromethyl)aniline
N-[2-(4-Ethylphenoxy)propyl]-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₈H₂₀F₃NO
MDL Number: MFCD10687505
CAS Number: 1040685-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR