N-(4-Propoxybenzyl)-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

031379-500MG
101831-584EA 306.15 USD
101831-584
N-(4-Propoxybenzyl)-3-(trifluoromethyl)aniline
N-(4-Propoxybenzyl)-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₇H₁₈F₃NO
MDL Number: MFCD10687504
CAS Number: 1040685-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR