N-(3-Chlorobenzyl)-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

031377-500MG
101831-580EA 306.15 USD
101831-580
N-(3-Chlorobenzyl)-3-(trifluoromethyl)aniline
N-(3-Chlorobenzyl)-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₁ClF₃N
MDL Number: MFCD06408417
CAS Number: 1024319-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR