N-[4-(2,4-Dichlorophenoxy)butyl]-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

031374-500MG
101831-574EA 306.15 USD
101831-574
N-[4-(2,4-Dichlorophenoxy)butyl]-3-(trifluoromethyl)aniline
N-[4-(2,4-Dichlorophenoxy)butyl]-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₇H₁₆Cl₂F₃NO
MDL Number: MFCD10687501
CAS Number: 1040684-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR