(2-Chlorophenyl)-N-[2-(2-methoxyethoxy)benzyl]methanamine

Supplier: Matrix Scientific

031373-500MG
101831-572EA 306.15 USD
101831-572
(2-Chlorophenyl)-N-[2-(2-methoxyethoxy)benzyl]methanamine
(2-Chlorophenyl)-N-[2-(2-methoxyethoxy)benzyl]methanamine

Hazard: Irritant

Formula: C₁₇H₂₀ClNO₂
MDL Number: MFCD10687500
CAS Number: 1040684-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR