(2-Chlorophenyl)-N-(4-isobutoxybenzyl)methanamine

Supplier: Matrix Scientific

031370-500MG
101831-566EA 306.15 USD
101831-566
(2-Chlorophenyl)-N-(4-isobutoxybenzyl)methanamine
(2-Chlorophenyl)-N-(4-isobutoxybenzyl)methanamine

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD10687497
CAS Number: 1040684-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR