(2-Chlorophenyl)-N-(2-methoxybenzyl)methanamine

Supplier: Matrix Scientific

031364-500MG
101831-554EA 306.15 USD
101831-554
(2-Chlorophenyl)-N-(2-methoxybenzyl)methanamine
(2-Chlorophenyl)-N-(2-methoxybenzyl)methanamine

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD04555653
CAS Number: 861225-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR