N-(2-Chlorobenzyl)-4-(2,4-dichlorophenoxy)-1-butanamine

Supplier: Matrix Scientific

031361-500MG
101831-548EA 306.15 USD
101831-548
N-(2-Chlorobenzyl)-4-(2,4-dichlorophenoxy)-1-butanamine
N-(2-Chlorobenzyl)-4-(2,4-dichlorophenoxy)-1-butanamine

Hazard: Irritant

Formula: C₁₇H₁₈Cl₃NO
MDL Number: MFCD10687490
CAS Number: 1040684-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR