N-(2-Furylmethyl)-2-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

031353-500MG
101831-532EA 306.15 USD
101831-532
N-(2-Furylmethyl)-2-(tetrahydro-2-furanylmethoxy)aniline
N-(2-Furylmethyl)-2-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₁₆H₁₉NO₃
MW: 273.33 g/mol
MDL Number: MFCD10687483
CAS Number: 1040686-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR