N-[3-(2-Chlorophenyl)propyl]-3-(2-methoxyethoxy)aniline

Supplier: Matrix Scientific

031351-500MG
101831-528EA 306.15 USD
101831-528
N-[3-(2-Chlorophenyl)propyl]-3-(2-methoxyethoxy)aniline
N-[3-(2-Chlorophenyl)propyl]-3-(2-methoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO₂
MW: 319.83 g/mol
MDL Number: MFCD10687481
CAS Number: 1040686-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR