N-(3,5-Dichlorobenzyl)-3-(2-methoxyethoxy)aniline

Supplier: Matrix Scientific

031349-500MG
101831-524EA 306.15 USD
101831-524
N-(3,5-Dichlorobenzyl)-3-(2-methoxyethoxy)aniline
N-(3,5-Dichlorobenzyl)-3-(2-methoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO₂
MDL Number: MFCD10687479
CAS Number: 1040686-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR