N-[2-(2-Ethoxyethoxy)benzyl]-3-(2-methoxyethoxy)aniline

Supplier: Matrix Scientific

031348-500MG
101831-522EA 306.15 USD
101831-522
N-[2-(2-Ethoxyethoxy)benzyl]-3-(2-methoxyethoxy)aniline
N-[2-(2-Ethoxyethoxy)benzyl]-3-(2-methoxyethoxy)aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₄
MDL Number: MFCD10687478
CAS Number: 1040686-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR