N-[3-(2-Methoxyethoxy)phenyl]-N-[2-(3-methylphenoxy)propyl]amine

Supplier: Matrix Scientific

031342-500MG
101831-510EA 306.15 USD
101831-510
N-[3-(2-Methoxyethoxy)phenyl]-N-[2-(3-methylphenoxy)propyl]amine
N-[3-(2-Methoxyethoxy)phenyl]-N-[2-(3-methylphenoxy)propyl]amine

Hazard: Irritant

Formula: C₁₉H₂₅NO₃
MDL Number: MFCD10687473
CAS Number: 1040686-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR