N-(2,4-Dichlorobenzyl)-3-(2-methoxyethoxy)aniline

Supplier: Matrix Scientific

031339-500MG
101831-504EA 306.15 USD
101831-504
N-(2,4-Dichlorobenzyl)-3-(2-methoxyethoxy)aniline
N-(2,4-Dichlorobenzyl)-3-(2-methoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO₂
MDL Number: MFCD10687470
CAS Number: 1040686-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR