3-(sec-Butoxy)-N-(2-isopropoxybenzyl)aniline

Supplier: Matrix Scientific

031336-500MG
101831-498EA 306.15 USD
101831-498
3-(sec-Butoxy)-N-(2-isopropoxybenzyl)aniline
3-(sec-Butoxy)-N-(2-isopropoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₀H₂₇NO₂
MDL Number: MFCD10687467
CAS Number: 1040686-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR