3-(sec-Butoxy)-N-[2-(hexyloxy)benzyl]aniline

Supplier: Matrix Scientific

031334-500MG
101831-494EA 306.15 USD
101831-494
3-(sec-Butoxy)-N-[2-(hexyloxy)benzyl]aniline
3-(sec-Butoxy)-N-[2-(hexyloxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₃H₃₃NO₂
MDL Number: MFCD10687465
CAS Number: 1040685-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR