N-[3-(sec-Butoxy)phenyl]-N-[2-(2-chlorophenoxy)propyl]amine

Supplier: Matrix Scientific

031332-500MG
101831-490EA 306.15 USD
101831-490
N-[3-(sec-Butoxy)phenyl]-N-[2-(2-chlorophenoxy)propyl]amine
N-[3-(sec-Butoxy)phenyl]-N-[2-(2-chlorophenoxy)propyl]amine

Hazard: Irritant

Formula: C₁₉H₂₄ClNO₂
MW: 333.86 g/mol
MDL Number: MFCD10687463
CAS Number: 1040685-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR