N-[2-(2,6-Dimethylphenoxy)ethyl]-2,4-dimethylaniline
Supplier: Matrix Scientific
031331-500MG
101831-488EA
306.15
USD
101831-488
N-[2-(2,6-Dimethylphenoxy)ethyl]-2,4-dimethylaniline
N-[2-(2,6-Dimethylphenoxy)ethyl]-2,4-dimethylaniline
Hazard: Irritant
Formula:
C₁₈H₂₃NO |
MDL Number:
MFCD10687462 CAS Number: 1040685-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...