3-(sec-Butoxy)-N-(3-methylbenzyl)aniline

Supplier: Matrix Scientific

031329-500MG
101831-484EA 306.15 USD
101831-484
3-(sec-Butoxy)-N-(3-methylbenzyl)aniline
3-(sec-Butoxy)-N-(3-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10687460
CAS Number: 1040685-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR