3-(sec-Butoxy)-N-(2-methylbenzyl)aniline

Supplier: Matrix Scientific

031328-500MG
101831-482EA 306.15 USD
101831-482
3-(sec-Butoxy)-N-(2-methylbenzyl)aniline
3-(sec-Butoxy)-N-(2-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10687459
CAS Number: 1040685-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR