N-[4-(sec-Butoxy)phenyl]-N-[2-(2-phenoxyethoxy)benzyl]amine

Supplier: Matrix Scientific

031325-500MG
101831-476EA 306.15 USD
101831-476
N-[4-(sec-Butoxy)phenyl]-N-[2-(2-phenoxyethoxy)benzyl]amine
N-[4-(sec-Butoxy)phenyl]-N-[2-(2-phenoxyethoxy)benzyl]amine

Hazard: Irritant

Formula: C₂₅H₂₉NO₃
MW: 391.51 g/mol
MDL Number: MFCD10687456
CAS Number: 1040685-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR