4-(sec-Butoxy)-N-(4-isobutoxybenzyl)aniline

Supplier: Matrix Scientific

031323-500MG
101831-472EA 306.15 USD
101831-472
4-(sec-Butoxy)-N-(4-isobutoxybenzyl)aniline
4-(sec-Butoxy)-N-(4-isobutoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10687454
CAS Number: 1040685-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR