4-(sec-Butoxy)-N-(4-butoxybenzyl)aniline

Supplier: Matrix Scientific

031322-500MG
101831-470EA 306.15 USD
101831-470
4-(sec-Butoxy)-N-(4-butoxybenzyl)aniline
4-(sec-Butoxy)-N-(4-butoxybenzyl)aniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10687453
CAS Number: 1040685-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR